
8-Aminoclonazolam solution

Synonym(s):
8-Aminoclonazolam solution
Empirical Formula (Hill Notation):
C17H14ClN5
CAS Number:
Molecular Weight:
323.78
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing