
4-Methylumbelliferyl N-acetyl-β-D-glucosaminide

Synonym(s):
4-Methylumbelliferyl-2-acetamido-2-deoxy-β-D-glucopyranoside, 4-Methylumbelliferyl-2-acetamido-2-deoxy-β-D-glucopyranoside, 4-Methylumbelliferyl-N-acetyl-β-D-glucosaminide
Empirical Formula (Hill Notation):
C18H21NO8
CAS Number:
Molecular Weight:
379.36
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-3 of 3
Compare
Product Number
Description
Pricing