
4-ANPP solution

Empirical Formula (Hill Notation):
C19H24N2
CAS Number:
Molecular Weight:
280.41
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing