
3,6-Dibromocarbazole

Empirical Formula (Hill Notation):
C12H7Br2N
CAS Number:
Molecular Weight:
325.00
Products
Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing