Skip to Content
Merck
2-[4-[(7-Chloro-4-quinolinyl)amino]pentyl] amino ethanol British Pharmacopoeia (BP) Reference Standard

2-[4-[(7-Chloro-4-quinolinyl)amino]pentyl] amino ethanol

2-[4-[(7-Chloro-4-quinolinyl)amino]pentyl] amino ethanol British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Hydroxychloroquine sulfate, 7-Chloro-4-[4-(N-ethyl-N-b-hydroxyethylamino)-1-methylbutylamino]quinoline sulfate
Empirical Formula (Hill Notation):
C18H26ClN3O · H2SO4
CAS Number:
Molecular Weight:
433.95

Products

Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1