
(1S)-(−)-Camphorsulfonylimine

Synonym(s):
(1S)-(−)-10,10-Dimethyl-3-thia-4-azatricyclo[5.2.1.01,5]dec-4-ene 3,3-dioxide, (3aS)-(−)-4,5,6,7-Tetrahydro-8,8-dimethyl-3H-3a,6-methano-2,1-benzisothiazole-2,2-dioxide
Empirical Formula (Hill Notation):
C10H15NO2S
CAS Number:
Molecular Weight:
213.30
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing