Skip to Content
Merck
All Photos(1)

Key Documents

FTC-236

6030202, human thyroid (lymph node metastasis), Endothelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

FTC-236, 06030202

biological source

human thyroid (lymph node metastasis)

growth mode

Adherent

karyotype

Not specified

morphology

Endothelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Thyroid Cancer Neck Lymph Node Metastasis

Cell Line Description

Derived from a neck lymph node metastasis of a follicular thyroid cancer (FTC).This cell line is tumourigenic in Balb/c mice. FTC-133 (Sigma Catalogue number 94060901), FTC-236 (Sigma Catalogue number 06030202) and FTC-238 (Sigma Catalogue number 94060902) were all derived from the same individual, a 42 year old male with follicular thyroid cancer. This has been confirmed by short tandem repeat STR-PCR analysis at ECACC The Y chromosome could not be detected in these cell line by (STR)-PCR analysis. It is a known phenomenon that due to the increased genetic instability of cancer cell lines the Y chromosome can be rearranged or lost resulting in lack of detection. The cell lines are identical to the source provided by the depositor based on the STR-PCR analysis.

Culture Medium

DMEM and Ham′s F12 (1:1 mixture) + 10ug/ml Insulin + 0.01U/ml Thyroid Stimulating Hormone (TSH) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service