8.21052
2,4-Pyridinedicarboxylic acid
for synthesis
Synonym(s):
2,4-Pyridinedicarboxylic acid, Lutidinic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
form
solid
mp
242-243 °C
solubility
2.49 g/L
storage temp.
2-30°C
InChI
1S/C7H5NO4/c9-6(10)4-1-2-8-5(3-4)7(11)12/h1-3H,(H,9,10)(H,11,12)
InChI key
MJIVRKPEXXHNJT-UHFFFAOYSA-N
Analysis Note
Assay (acidimetric, calc. on anhydrous substance): ≥ 98.0 %
Water (K. F.): ≤ 1.0 %
Identity (IR): passes test
Water (K. F.): ≤ 1.0 %
Identity (IR): passes test
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service