GF53360614
Polyamide - Nylon 6
(PA 6), monofilament, diam. 1 mm, L 50 m, condition nylon 6/66 copolymer
Synonym(s):
AM305990
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
monofilament
manufacturer/tradename
Goodfellow 533-606-14
L
50 m
diam.
1 mm
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service