HPLC Analysis Carbohydrates on Proteomix® SAX-NP5
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Proteomix SAX-NP5, 15 cm x 4.6 mm I.D., 5 μm particles (Z777228)
mobile phase
[A] 0.05% (25% ammonium hydroxide) in acetonitrile; [B] 0.05% (25% ammonium hydroxide) in water
gradient
15 to 80% B in 20 min
flow rate
0.7 mL/min
temp.
35 °C
detector
ELSD
sample
Corn starch hydrolysate; glucose oligomer (G1 to G6)
Description
Legal Information
Proteomix is a registered trademark of Sepax Technologies