Merck
  • Effects of chilled-then-frozen storage (up to 52weeks) on lamb M. longissimus lumborum quality and safety parameters.
Effects of chilled-then-frozen storage (up to 52weeks) on lamb M. longissimus lumborum quality and safety parameters.
Meat science (2017-08-05)
Cassius E O Coombs, Benjamin W B Holman, Damian Collins, Michael A Friend, David L Hopkins
ABSTRACT
This study evaluated the effect of chilled followed by frozen storage on lamb quality and safety parameters. Experimental (n=360) M. longissimus lumborum (LL) were randomly sampled from the boning room of a commercial Australian abattoir, at 24 h post-mortem, and assigned to five chilled storage periods (0, 2, 4, 6 and 8 weeks) and six subsequent frozen storage periods (0, 4, 8, 12, 24 and 52 weeks). Upon completion of each storage treatment combination, corresponding LL were sub-sectioned and analysed for colour stability (0, 1, 2 and 3 days), shear force, fluid losses (purge, thaw and cooking losses), intramuscular fat content, sarcomere length, water activity and microbial load (lactic acid bacteria, Enterobacteriaceae sp., Brochothrix thermosphacta, Clostridium perfringens and Escherichia coli). LL stored chilled for 2-4 weeks prior to freezing presented superior results for shear force, display colour and low levels of spoilage microbes, correlating with good eating quality and safety following more than one year of frozen storage.