Merck
  • Synchronous fluorescence spectrometry of 1-hydroxypyrene: a rapid screening method for identification of PAH exposure in tissue from marine polychaetes.
Synchronous fluorescence spectrometry of 1-hydroxypyrene: a rapid screening method for identification of PAH exposure in tissue from marine polychaetes.
Marine environmental research (2003-08-21)
Anders M B Giessing, Lawrence M Mayer, Thomas L Forbes
ABSTRACT
The uptake of polycyclic aromatic hydrocarbons (PAHs) by marine deposit-feeding invertebrates can be determined by screening for PAH-derived metabolites. We identified 1-hydroxypyrene as the only intermediate metabolite in tissue of four species of deposit-feeding polychaetes, Nereis diversicolor, Nereis virens, Arenicola marina, and Capitella sp. I exposed to pyrene spiked sediment. Synchronous fluorescence spectroscopy (SFS) provides a fast and simple method for both qualitative and quantitative analysis of 1-hydroxypyrene in all four species. The SFS assay was validated using HPLC with ultraviolet detection. A good correlation between 1-hydroxypyrene concentrations determined by the two methods was observed. We used HPLC with fluorescence detection combined with enzymatic hydrolysis of conjugated metabolites to investigate species specific metabolite patterns. A tentative aqueous metabolite identification scheme indicates that Nereid polychaetes predominately make use of glucuronide conjugation whereas Capitella sp. I. and Arenicola marina appear to utilize predominantly sulfate and/or glucoside conjugation. The usefulness of 1-hydroxypyrene as a biomarker for PAH exposure in deposit-feeding invertebrates is discussed.
Sigma-Aldrich
1-Hydroxypyrene, 98%