Merck
  • Zn(II), Ni(II), Cu(II) and Pb(II) complexes of tridentate asymmetrical Schiff base ligands: synthesis, characterization, properties and biological activity.
Zn(II), Ni(II), Cu(II) and Pb(II) complexes of tridentate asymmetrical Schiff base ligands: synthesis, characterization, properties and biological activity.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-01-01)
Mustafa Şahin, Nuriye Koçak, Damla Erdenay, Uğur Arslan
ABSTRACT
New asymmetrical tridentate Schiff base ligands were synthesized using 1,2-phenylenediamine, 4-methyl-1,2-phenylenediamine, 2-hydroxy-1-napthaldehyde, 9-anthracenecarboxaldehyde. Schiff base ligands and their metal complexes were synthesised and characterized by using FT-IR, (1)H NMR, (13)C NMR, UV-Vis, XRD, ESR, elemental analysis and fluorescence studies. The antimicrobial activity of the ligands and their metal complexes were studied against Staphylococcus aureus ATCC 29213, S. aureus ATCC 25923, Streptococcus mutans RSHM 676, Enterococcus faecalis ATCC 29212, Escherichia coli ATCC 25922, Pseudomonas aeruginosa ATCC 27853. The determination of the antibacterial activity was done using the broth microdilution methods. In general, it has been determined that the studied compounds have MIC values similar to Gram-positive and Gram-negative bacteria. It has been found that Ni, Pb, Zn derivatives of HL1A and ZnL(2)A has lower MIC values than ampicillin for P. aeruginosa ATCC 27853 strain.
Sigma-Aldrich
9-Anthracenecarboxaldehyde, 97%
Pricing and availability is not currently available.