Merck
  • A quantitative liposome microarray to systematically characterize protein-lipid interactions.
A quantitative liposome microarray to systematically characterize protein-lipid interactions.
Nature methods (2013-11-26)
Antoine-Emmanuel Saliba, Ivana Vonkova, Stefano Ceschia, Greg M Findlay, Kenji Maeda, Christian Tischer, Samy Deghou, Vera van Noort, Peer Bork, Tony Pawson, Jan Ellenberg, Anne-Claude Gavin
ABSTRACT
Lipids have a role in virtually all biological processes, acting as structural elements, scaffolds and signaling molecules, but they are still largely under-represented in known biological networks. Here we describe a liposome microarray-based assay (LiMA), a method that measures protein recruitment to membranes in a quantitative, automated, multiplexed and high-throughput manner.
Sigma-Aldrich
Agarose, Type IV, Special High EEO
Pricing and availability is not currently available.
Sigma-Aldrich
Agarose, High Gelling Temperature
Pricing and availability is not currently available.
Sigma-Aldrich
Sepharose 6B, 6% Beaded Agarose, 45-165 μm (wet), fractionation range 10,000-1,000,000 Da (dextrans)
Pricing and availability is not currently available.
Sigma-Aldrich
Agarose, For pulsed field electrophoresis running gel
Pricing and availability is not currently available.
Sigma-Aldrich
Agarose, Low EEO
Pricing and availability is not currently available.
Sigma-Aldrich
Agarose, Type II-A, Medium EEO
Pricing and availability is not currently available.
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature
Pricing and availability is not currently available.
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature, molecular biology grade
Pricing and availability is not currently available.
Sigma-Aldrich
Agarose, Type I, low EEO
Pricing and availability is not currently available.
Sigma-Aldrich
Agarose, Special High EEO
Pricing and availability is not currently available.
Sigma-Aldrich
Agarose, Type I-A, low EEO
Pricing and availability is not currently available.
Sigma-Aldrich
Agarose, Low EEO, for Immunoelectrophoresis
Pricing and availability is not currently available.
Sigma-Aldrich
Agarose, Wide range, for molecular biology
Pricing and availability is not currently available.
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, Wide range/Standard 3:1
Pricing and availability is not currently available.
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, low EEO
Pricing and availability is not currently available.
Sigma-Aldrich
Agarose, Medium EEO, for molecular biology
Pricing and availability is not currently available.
Sigma-Aldrich
Agarose, for molecular biology
Pricing and availability is not currently available.
Sigma-Aldrich
Agarose, High EEO, for molecular biology
Pricing and availability is not currently available.
Sigma-Aldrich
Agarose, High EEO
Pricing and availability is not currently available.
Sigma-Aldrich
Sepharose 4B, 45-165 μm bead diameter
Pricing and availability is not currently available.