Merck
  • [Induction of micronuclei in polychromatic erythrocytes from mouse bone marrow following intraperitoneal administration of halogenated benzenes].
[Induction of micronuclei in polychromatic erythrocytes from mouse bone marrow following intraperitoneal administration of halogenated benzenes].
Bollettino della Societa italiana di biologia sperimentale (1990-07-01)
M Parrini, C Bolognesi, P Roggieri
ABSTRACT
The mutagenicity of halogenated benzenes, including three isomers of tri- and tetrachlorobenzenes (TCB, TeCB) was studied on male Swiss CD1 mice by MN test. The data presented show that all the halogenated benzenes tested were found to be clastogenic apart from 1,2,3,5-TeCB. No significant differences were observed in the clastogenic activities of TCB-isomers and TeCB-isomers.
Product Description
Supelco
1,2,3,5-Tetrachlorobenzene, PESTANAL®, analytical standard