Merck
  • The protective role of glutathione peroxidase in apoptosis induced by reactive oxygen species.
The protective role of glutathione peroxidase in apoptosis induced by reactive oxygen species.
Journal of biochemistry (1996-04-01)
Y Kayanoki, J Fujii, K N Islam, K Suzuki, S Kawata, Y Matsuzawa, N Taniguchi
ABSTRACT
Selenium-dependent glutathione peroxidase (GPx) plays a protective role in oxidative stress-induced apoptosis. In this study, we demonstrated that MDBK cells, a bovine renal epithelial cell line, exhibited internucleosomal DNA fragmentation characteristic of apoptotic cell death under selenium-deficient conditions with lower doses of hydrogen peroxide (H2O2) than under selenium-supplemented ones. This was due to a decreased amount of GPx in the cells under selenium-deficient conditions, because other antioxidative enzyme activities were not affected by the selenium supplementation. Cumene hydroperoxide also induced DNA fragmentation in selenium-deficient cells but no ladder formation was observed. Flow cytometric analysis showed that selenium-deficient cells were less capable of scavenging intracellular peroxides after exposure to exogenous H2O2 than selenium-supplemented ones. In contrast, there was no difference in viability between selenium-supplemented and non-supplemented cells in cell survival after exposure to menadione, which activates the electron transport system and increases intracellular superoxide radicals. Clofibrate, a peroxisomal proliferator and an inducer of catalase (CAT), partially protected both Se-deficient and Se-supplemented cells from exogenous H202. We concluded that selenium-deficient cells were more easily brought to apoptotic cell death by peroxides, but not by superoxide radicals, than selenium-supplemented ones and that CAT could compensate for the depletion of GPx to a certain degree by scavenging H2O2.
Sigma-Aldrich
Selenous acid, 98%
Sigma-Aldrich
Selenous acid, 99.999% trace metals basis