Merck
  • Simultaneous determination of aminopyrine hydroxylation and aminopyrine N-demethylation in liver microsomes by high-performance liquid chromatography.
Simultaneous determination of aminopyrine hydroxylation and aminopyrine N-demethylation in liver microsomes by high-performance liquid chromatography.
Journal of chromatography (1983-05-13)
K Inoue, K Fujimori, K Mizokami, M Sunouchi, A Takanaka, Y Omori
ABSTRACT
Aminopyrine and its metabolites, including 3-hydroxymethyl-2-methyl-4-dimethylamino-1-phenyl-3-pyrazoline-5-one which is a hydroxylated metabolite of aminopyrine, were separated on a reversed-phase (C8) Radial-Pak column using a mobile phase of methanol-triethylamine-water (30:1:69) adjusted to pH 5.40 with acetic acid. Detection of the peak was performed by an ultraviolet detector at 254 nm. By the rapid and simple method, aminopyrine hydroxylation as well as aminopyrine N-demethylation in liver microsomes can be examined simultaneously.
Product Description
Supelco
4-Formylaminoantipyrine, VETRANAL®, analytical standard
Metamizole impurity A, European Pharmacopoeia (EP) Reference Standard