Merck
Role of GABA in the control of thyrotropin secretion in the rat.
Brain research (1983-05-23)
D Jordan, C Poncet, M Veisseire, R Mornex
ABSTRACT
Various doses of GABA from 0.25 to 5 mumol injected into the third ventricle decrease serum TSH rapidly. The same effect was observed with GABOB (10 mumol), the hydroxylated form of GABA. The inhibitory effect of both of these drugs was prevented by picrotoxin injection (1 microgram). Peripheral injection of GABAergic drugs such as GABOB or AOAA also decreased serum TSH. In vitro, the addition of GABA (from 6.7 . 10(-6) to 6.7 . 10(-4) M) to the incubation medium of hemi-anterior pituitary did not modify the liberation of TSH. To test the physiological role of GABA in the regulation of thyrotropin function the circadian TSH rhythm was used as a model. Both GABAergic inhibitors, picrotoxin (10 microgram/kg b.w.) as well as semicarbazide (150 mg/kg b.w.), induced an increase of the low basal nocturnal level of TSH (centered on the 02.00 h time point) without altering the diurnal peak of TSH. We conclude that GABA has an inhibitory effect on central thyrotropin control via an inhibition of TRH release from the hypothalamus and might be, at least partly, responsible for the low nocturnal levels of serum TSH observed during the circadian physiological rhythm.
Sigma-Aldrich
4-Amino-3-hydroxybutyric acid, 98%
Sigma-Aldrich
(S)-(+)-4-Amino-3-hydroxybutyric acid, 97%