Merck
  • Effects of alpha-tocopherol on the lipid peroxidation and fluidity of porcine intestinal brush-border membranes.
Effects of alpha-tocopherol on the lipid peroxidation and fluidity of porcine intestinal brush-border membranes.
Biochimica et biophysica acta (1986-06-26)
T Ohyashiki, H Ushiro, T Mohri
ABSTRACT
The effect of alpha-tocopherol on the lipid fluidity of porcine intestinal brush-border membranes was studied using pyrene as a fluorescent probe. Addition of alpha-tocopherol to the medium decreased fluorescence intensity and lifetime, but increased the fluorescence polarization of pyrene-labeled membranes. beta-, gamma-, and delta-Tocopherols gave no appreciable effect on the fluorescence intensity and polarization of the complex. The apparent dissociation constant (3.1 +/- 0.12 microM) of the interaction of alpha-tocopherol with the membranes, estimated from the change in the fluorescence intensity with varying concentrations of alpha-tocopherol, was in good agreement with the concentration required to cause the half-maximal inhibition of lipid peroxidation of the membranes performed by incubation with 100 microM ascorbic acid and 10 microM Fe2+. Decrease of the slope in the thermal Perrin plot of the polarization of pyrene-labeled membranes by alpha-tocopherol suggests that the movement of pyrene molecules in the membranes is restricted by binding of the tocopherol. This interpretation was confirmed by an increased harmonic mean of the rotational relaxation time of the dye molecules in the membranes from 10.9 +/- 0.16 to 18.5 +/- 0.51 microseconds after addition of 25 microM alpha-tocopherol to the medium. The perturbation of lipid phase in the membranes induced by alpha-tocopherol was also suggested from a decreased quenching rate constant of pyrene fluorescence in the membranes for Tl+. Based on these results, the effect of alpha-tocopherol on the lipid fluidity of the membranes is discussed.
Sigma-Aldrich
Thallium(I) acetate, ≥99%
Sigma-Aldrich
Thallium(I) acetate, purum, ≥97.0%