Merck
  • A comparison of CMV detection in gastrointestinal mucosal biopsies using immunohistochemistry and PCR performed on formalin-fixed, paraffin-embedded tissue.
A comparison of CMV detection in gastrointestinal mucosal biopsies using immunohistochemistry and PCR performed on formalin-fixed, paraffin-embedded tissue.
The American journal of surgical pathology (2013-05-08)
Anne M Mills, Frances P Guo, Andrew P Copland, Reetesh K Pai, Benjamin A Pinsky
ABSTRACT
Cytomegalovirus (CMV) can precipitate and exacerbate gastrointestinal (GI) mucosal injury. The gold standard for CMV detection in formalin-fixed, paraffin-embedded (FFPE) tissue is immunohistochemistry (IHC). Although CMV polymerase chain reaction (PCR) on fresh tissue may be a valuable adjunct to IHC, its utility is unknown for FFPE tissues. We therefore evaluated quantitative, real-time CMV PCR in a total of 102 FFPE GI biopsy specimens from 74 patients with a history of hematopoietic stem cell or solid organ transplant, inflammatory bowel disease, human immunodeficiency virus infection, or unspecified colitis. CMV DNA was detected by PCR in 90.9% (30/33) of IHC-positive, 14.5% (8/55) of IHC-negative, and 20.0% (1/5) of IHC-equivocal FFPE tissues. Quantitation of CMV DNA copies normalized to β-globin demonstrated a wide range of values (median 0.276; range, 0.0004 to 144.50). Importantly, 93.3% (14/15) of patients with IHC-positive, active colitis showed no evidence of CMV in matched concurrent, histologically normal biopsies tested by PCR. These results suggest that CMV PCR on FFPE GI biopsies complements IHC and has the potential to identify additional patients who may benefit from anti-CMV therapy.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Formaldehyde neutral buffer solution, 3.7% formaldehyde in H2O
Sigma-Aldrich
Formaldehyde neutral buffer solution, 7.5% formaldehyde in H2O
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O