Merck
  • Characterization of changes of lignin structure in the processes of cooking with solid alkali and different active oxygen.
Characterization of changes of lignin structure in the processes of cooking with solid alkali and different active oxygen.
Bioresource technology (2012-09-04)
Qiulin Yang, Jianbin Shi, Lu Lin, Lincai Peng, Junping Zhuang
ABSTRACT
The cooking with solid alkali and active oxygen has a high selectivity for delignification. In the present work, the O(2) and H(2)O(2) were separately combined with MgO used in cornstalk cooking for investigating their effects on delignification. After cooking, the lignins in raw material, pulp, and yellow liquor were all characterized by HSQC NMR. The results showed that the syringyl (S/S'/S″) units and β-O-4' (A/A'/A″) structures had different reactivity in the cooking with MgO and H(2)O(2) due to their different structures on side-chains. Whereas the syringyl (S/S'/S″) units could be completely decomposed when the MgO and O(2) were used, and the β-O-4' (A/A'/A″) structures could be partly degraded. A novel structure G' unit with a carbonyl group was only generated in the cooking with MgO and O(2). In addition, the H unit, non-phenolic β-β' (B) and β-5' (C) structures were all stable in both of the two cooking processes.
Sigma-Aldrich
Magnesium oxide, 99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, nanopowder, ≤50 nm particle size (BET)
Sigma-Aldrich
Magnesium oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, BioUltra, ≥97.0% (calcined substance, KT)
Sigma-Aldrich
Magnesium oxide, tested according to Ph. Eur., heavy
Sigma-Aldrich
Magnesium oxide, light, 95%
Sigma-Aldrich
Magnesium oxide, -10-+50 mesh, 98%
Sigma-Aldrich
Magnesium oxide, ACS reagent, 97%
Sigma-Aldrich
Magnesium oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 530, light, 98.0-100.5% (calc. for dried substance)
Sigma-Aldrich
Magnesium oxide, ≥99% trace metals basis, -325 mesh
Sigma-Aldrich
Magnesium oxide, (single crystal substrate), ≥99.9% trace metals basis, <100>, L × W × thickness 10 mm × 10 mm × 0.5 mm