Merck
  • Systematic review: racecadotril in the treatment of acute diarrhoea in children.
Systematic review: racecadotril in the treatment of acute diarrhoea in children.
Alimentary pharmacology & therapeutics (2007-09-05)
H Szajewska, M Ruszczyński, A Chmielewska, J Wieczorek
ABSTRACT
Racecadotril (acetorphan) is an antisecretory drug that exerts its antidiarrhoeal effects by inhibiting intestinal enkephalinase. To summarize studies testing the efficacy and safety of racecadotril for treating children with acute gastroenteritis. Reports were gathered by searching electronic databases MEDLINE, EMBASE, the Cochrane Library (all up to April 2007), relevant journals, and bibliographies of reviewed articles. Only randomized-controlled trials were included. Three randomized-controlled trials (471 participants) met the inclusion criteria. Two trials reported stool output, and data suggested less stool output in the racecadotril group than in the control group. The duration of diarrhoea was significantly reduced in the three trials reporting this outcome. Achievement of a cure by day 5 was similar in both groups. Adverse effects were similar in both groups. The small number of included trials provided some evidence in favour of the use of racecadotril over placebo or no intervention, to reduce the stool output and duration of diarrhoea in children with acute gastroenteritis. However, more data in out-patients are needed. The safety as well as the cost-effectiveness of the therapy should be explored, before routine therapy with racecadotril is recommended.
Product Description
Racecadotril for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Racecadotril, ≥98% (HPLC)
Racecadotril, European Pharmacopoeia (EP) Reference Standard