Merck
  • Individual NMDA receptor GluN2 subunit signaling domains differentially regulate the postnatal maturation of hippocampal excitatory synaptic transmission and plasticity but not dendritic morphology.
Individual NMDA receptor GluN2 subunit signaling domains differentially regulate the postnatal maturation of hippocampal excitatory synaptic transmission and plasticity but not dendritic morphology.
Synapse (New York, N.Y.) (2024-05-30)
Rachel E Keith, Grace A Wild, Matthew J Keith, Diyi Chen, Svetlana Pack, Theodore C Dumas
ABSTRACT
N-methyl-d-aspartate receptors (NMDARs) at hippocampal excitatory synapses undergo a late postnatal shift in subunit composition, from an initial prevalence of GluN2B subunit incorporation to a later predominance of GluN2A. This GluN2B to GluN2A shift alters NMDAR calcium conductance dynamics and intracellular molecular signaling that are individually regulated by distinct GluN2 signaling domains and temporally align with developmental alterations in dendritic and synaptic plasticity. However, the impacts of individual GluN2B to GluN2A signaling domains on neuronal development remain unknown. Ionotropic and intracellular signaling domains of GluN2 subunits were separated by creating chimeric GluN2 subunits that were expressed in two transgenic mouse lines. Western blot and immunoprecipitation revealed that roughly one third of native synaptic NMDARs were replaced by transformed NMDARs without altering total synaptic NMDAR content. Schaffer collateral synaptic strength was transiently increased in acutely prepared hippocampal slices at just over 3 weeks of age in animals overexpressing the GluN2B carboxy terminus. Long-term potentiation (LTP) induction following lower frequency stimulation was regulated by GluN2 ionotropic signaling domains in an age-dependent manner and LTP maintenance was enhanced by overexpression of the GluN2B CTD in mature animals. After higher frequency stimulation, the induction and maintenance of LTP were increased in young adult animals overexpressing the GluN2B ionotropic signaling domains but reduced in juveniles just over 3 weeks of age. Confocal imaging of green fluorescent protein (GFP)- labeled CA1 pyramidal neurons revealed no alterations in dendritic morphology or spine density in mice expressing chimeric GluN2 subunits. These results illustrate how individual GluN2 subunit signaling domains do or do not control physiological and morphological development of hippocampal excitatory neurons and better clarify the neurobiological factors that govern hippocampal maturation. SIGNIFICANCE STATEMENT: A developmental reduction in the magnitude of hippocampal long-term synaptic potentiation (LTP) and a concomitant improvement in spatial maze performance coincide with greater incorporation of GluN2A subunits into synaptic NMDARs. Corroborating our prior discovery that overexpression of GluN2A-type ionotropic signaling domains enables context-based navigation in immature mice, GluN2A-type ionotropic signaling domain overexpression reduces LTP induction threshold and magnitude in immature mice. Also, we previously found that GluN2B carboxy terminal domain (CTD) overexpression enhances long-term spatial memory in mature mice and now report that the GluN2B CTD is associated with greater amplitude of LTP after induction in mature mice. Thus, the late postnatal maturation of context encoding likely relies on a shift toward GluN2A-type ionotropic signaling and a reduction in the threshold to induce LTP while memory consolidation and LTP maintenance are regulated by GluN2B subunit CTD signaling.
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, (H+L) Alkaline Phosphatase conjugate, 1 mg/mL, Chemicon®
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Alkaline Phosphatase conjugate, 0.6 mg/mL, Chemicon®