Merck
  • CDK1 bridges NF-κB and β-catenin signaling in response to H. pylori infection in gastric tumorigenesis.
CDK1 bridges NF-κB and β-catenin signaling in response to H. pylori infection in gastric tumorigenesis.
Cell reports (2023-01-23)
Shoumin Zhu, Marwah Al-Mathkour, Longlong Cao, Shayan Khalafi, Zheng Chen, Julio Poveda, Dunfa Peng, Heng Lu, Mohammed Soutto, Tianling Hu, Oliver G McDonald, Alexander Zaika, Wael El-Rifai
ABSTRACT
Infection with Helicobacter pylori (H. pylori) is the main risk factor for gastric cancer, a leading cause of cancer-related death worldwide. The oncogenic functions of cyclin-dependent kinase 1 (CDK1) are not fully understood in gastric tumorigenesis. Using public datasets, quantitative real-time PCR, western blot, and immunohistochemical (IHC) analyses, we detect high levels of CDK1 in human and mouse gastric tumors. H. pylori infection induces activation of nuclear factor κB (NF-κB) with a significant increase in CDK1 in in vitro and in vivo models (p < 0.01). We confirm active NF-κB binding sites on the CDK1 promoter sequence. CDK1 phosphorylates and inhibits GSK-3β activity through direct binding with subsequent accumulation and activation of β-catenin. CDK1 silencing or pharmacologic inhibition reverses these effects and impairs tumor organoids and spheroid formation. IHC analysis demonstrates a positive correlation between CDK1 and β-catenin. The results demonstrate a mechanistic link between infection, inflammation, and gastric tumorigenesis where CDK1 plays a critical role.
Sigma-Aldrich
Cdk1 Inhibitor IV, RO-3306, InSolution, ≥95%
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture