Merck
  • The p300 Inhibitor A-485 Exerts Antitumor Activity in Growth Hormone Pituitary Adenoma.
The p300 Inhibitor A-485 Exerts Antitumor Activity in Growth Hormone Pituitary Adenoma.
The Journal of clinical endocrinology and metabolism (2022-03-06)
Chenxing Ji, Wen Xu, Hong Ding, Zhengyuan Chen, Chengzhang Shi, Jie Han, Liang Yu, Nidan Qiao, Yichao Zhang, Xiaoyun Cao, Xiang Zhou, Haixia Cheng, Huijin Feng, Cheng Luo, Zhiyu Li, Bing Zhou, Zhao Ye, Yao Zhao
ABSTRACT
Growth hormone pituitary adenoma (GHPA), a major subtype of pituitary adenoma (PA), can lead to progressive somatic disfigurement, multiple complications, and even increased mortality. The efficacy of current treatments is limited; thus, a novel pharmacological treatment is urgently needed. As a histone acetyltransferase (HAT) coactivator, p300 can regulate the transcription of several genes that are crucial for PA tumorigenesis and progression. However, the role of p300 and its catalytic inhibitor in GHPA is still unclear. We aimed to identify the expression of p300 in GHPA and in normal pituitary glands. The expression of p300 was detected in GHPA and normal pituitary tissues. Genetic knockdown was performed by siRNA. The efficacy of the p300 inhibitor A-485 in the cell cycle, proliferation, apoptosis, and hormone secretion was investigated by flow cytometry, ELISAs, Western blotting, and qRT-PCR. RNA sequencing, bioinformatic analysis, and subsequent validation experiments were performed to reveal the potential biological mechanism of A-485. High expression of p300 was found in GHPA tissues compared with normal pituitary tissues. Knockdown of p300 inhibited cell proliferation and clone formation. Treatment with A-485 suppressed cell growth and inhibited the secretion of GH in vitro and in vivo. Further mechanistic studies showed that A-485 could downregulate the expression or activity of several oncogenes, such as genes in the Pttg1, c-Myc, cAMP and PI3K/AKT/mTOR signaling pathways, which are crucial for PA tumorigenesis and progression. Our findings demonstrate that inhibition of HAT p300 by its selective inhibitor A-485 is a promising therapy for GHPA.
Sigma-Aldrich
A-485, ≥98% (HPLC)