Merck
MLL1 is regulated by KSHV LANA and is important for virus latency.
Nucleic acids research (2021-12-02)
Min Tan, Shijun Li, Franceline Juillard, Rute Chitas, Tânia F Custódio, Han Xue, Agnieszka Szymula, Qiming Sun, Bing Liu, Ángel L Álvarez, She Chen, Jing Huang, J Pedro Simas, Colin E McVey, Kenneth M Kaye
ABSTRACT
Mixed lineage leukemia 1 (MLL1) is a histone methyltransferase. Kaposi's sarcoma-associated herpesvirus (KSHV) is a leading cause of malignancy in AIDS. KSHV latently infects tumor cells and its genome is decorated with epigenetic marks. Here, we show that KSHV latency-associated nuclear antigen (LANA) recruits MLL1 to viral DNA where it establishes H3K4me3 modifications at the extensive KSHV terminal repeat elements during primary infection. LANA interacts with MLL1 complex members, including WDR5, integrates into the MLL1 complex, and regulates MLL1 activity. We describe the 1.5-Å crystal structure of N-terminal LANA peptide complexed with MLL1 complex member WDR5, which reveals a potential regulatory mechanism. Disruption of MLL1 expression rendered KSHV latency establishment highly deficient. This deficiency was rescued by MLL1 but not by catalytically inactive MLL1. Therefore, MLL1 is LANA regulable and exerts a central role in virus infection. These results suggest broad potential for MLL1 regulation, including by non-host factors.
Sigma-Aldrich
Anti-HHV-8 LNA-1 Antibody, clone LN53, clone LN53, from rat
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid