Merck
  • Insufficient Iron Improves Pristane-Induced Lupus by Promoting Treg Cell Expansion.
Insufficient Iron Improves Pristane-Induced Lupus by Promoting Treg Cell Expansion.
Frontiers in immunology (2022-03-18)
Xiaofei Gao, Yang Song, Shuang Lu, Longyuan Hu, Meiling Zheng, Sujie Jia, Ming Zhao
ABSTRACT
Trace element iron affects T cell biology, but the knowledge about the role of iron in regulating Treg cell expansion is limited. Treg cells play an important role in keeping peripheral T cell tolerance, increasing Treg cell expansion is a promising therapeutic method for SLE. Here we showed that iron deficiency promotes Treg cell expansion by reducing ROS accumulation, improving the disease progression of pristane-induced lupus. Increased oxidative stress inhibits Treg cell differentiation by inducing cell apoptosis. Our data suggest that altering iron metabolism promotes Treg cell expansion by preventing oxidation-induced cell death, which may provide a potential therapeutic strategy for SLE.
Sigma-Aldrich
Anti-CD3 Mouse mAb (UCHT1), liquid, >95% (immunoglobulin, SDS-PAGE), clone UCHT1
Sigma-Aldrich
Anti-CD28 Mouse mAb (ANC28.1/5D10), liquid, >95% (immunoglobulin, SDS-PAGE), clone ANC28.1/5D10