Merck
Mechanically Watching the ClpXP Proteolytic Machinery.
Methods in molecular biology (Clifton, N.J.) (2016-11-16)
Juan Carlos Cordova, Adrian O Olivares, Matthew J Lang
ABSTRACT
Energy-dependent protein degradation is studied through the dual bead ClpXP motility assay. Processing of folded proteins involves recognition, unfolding, translocation, and degradation stages. A dual optical trap, in a passive force-clamp geometry, exhibits bead-to-bead displacements that directly follow subprocesses underlying protein degradation. Discrete nanometer-scale displacements of the bead position reveal steps, dwells and pauses during the unfolding and translocation substeps. With a few structural modifications to the protease machinery and an engineered substrate, the assay represents a "chassis" for the measurement of a wide range of substrates and related machinery. The methods described faithfully record our assay as implemented, including substrate design, wet assay preparation, and the motility assay experiment protocol. The strategies herein permit adaptation of the ClpXP mechanical assay to a wide range of protein degradation systems.
Sigma-Aldrich
Casein from bovine milk, technical grade
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type VII, lyophilized powder, ≥100,000 units/g solid (without added oxygen)
Sigma-Aldrich
Bovine Serum Albumin, OmniPur® Grade, >= 99%, Fraction V, Cold Alcohol Isolation