Merck
  • The relationships between systemic cytokine profiles and inflammatory markers in colorectal cancer and the prognostic significance of these parameters.
The relationships between systemic cytokine profiles and inflammatory markers in colorectal cancer and the prognostic significance of these parameters.
British journal of cancer (2020-06-04)
Ji Won Park, Hee Jin Chang, Hyun Yang Yeo, Nayoung Han, Byung Chang Kim, Sun-Young Kong, Jeongseon Kim, Jae Hwan Oh
ABSTRACT
Immunomodulatory cytokines and systemic inflammatory markers are important during cancer development and progression. This study investigated the association and prognostic impact of systemic cytokine profiles and inflammatory markers in colorectal cancer (CRC). Interleukin (IL)-1β, IL-6, IL-8, IL-9, IL-10, tumour necrosis factor (TNF)-α and vascular endothelial growth factor (VEGF) serum levels were measured using multiplex bead assays in CRC patients. Data on systemic inflammatory markers, such as the modified Glasgow prognostic score (mGPS), the neutrophil-to-lymphocyte ratio (NLR), platelet-to-lymphocyte ratio (PLR), lymphocyte-to-monocyte ratio (LMR), prognostic nutritional index (PNI) and fibrinogen, were collected. Survival analysis was performed to identify factors associated with progression-free survival (PFS) and overall survival (OS). There were moderate-to-strong correlations within serum cytokines, as well as within systemic inflammatory markers, whereas the associations between serum cytokines and systemic inflammatory markers were generally weak. IL-8 and the LMR were independent significant prognostic factors for PFS and OS. The low IL-8 and high LMR group had the best survival (both PFS and OS) of all groups. Systemic cytokine profiles and inflammatory markers have relatively weak intergroup correlations. A composite classification of systemic cytokine profiles and inflammatory markers has an enhanced prognostic value in CRC.
Sigma-Aldrich
Anti-HTR2B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-ZEB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution