Merck
  • The Relationship between the Structure and Properties of Amino Acid Ionic Liquids.
The Relationship between the Structure and Properties of Amino Acid Ionic Liquids.
Molecules (Basel, Switzerland) (2019-09-11)
Paula Ossowicz, Joanna Klebeko, Barbara Roman, Ewa Janus, Zbigniew Rozwadowski
ABSTRACT
Ionic liquids based on different l-amino acids (glycine, l-valine, l-leucine, l-isoleucine, l-histidine, l-methionine, l-tyrosine, l-tryptophan, l-arginine, and l-threonine) and different cations (tetrabutylammonium (TBA), tributylmethylammonium (tBMA), didecyldimethylammonium (DDA), (2-hydroxyethyl)trimethylammonium (choline) (Chol), alkyl(C12-C14) dimethylbenzylammonium (benzalkonium) (BA), dodecyltrimethylammonium (DDTMA), hexadecyltrimethylammonium (HDTMA), octadecyltrimethylammonium (ODTMA) and 1-ethyl-3-methylimidazolium (EMIM)) have been synthesized and characterized by NMR and FTIR. Viscosity, specific rotation, surface activity, thermal stability (TG), and phase transformations (DSC) have been determined and compared with available data. Furthermore, benzalkonium, didecyldimethylammonium, dodecyltrimethylammonium, hexadecyltrimethylammonium, and octadecyltrimethylammonium amino acid ionic liquids have been shown to exhibit surface activity. The dissolution of cellulose in amino acid ionic liquids (AAILs) composed of various cations was also investigated. Cellulose was only dissolved in EMIM salts of amino acids. In particular, the influence of the cation type on selected physicochemical and spectroscopic properties were discussed. The article is a mini review on amino acid ionic liquids.
Sigma-Aldrich
Trimethyloctadecylammonium chloride, ≥95.0% (calc. on dry substance, T)
Sigma-Aldrich
Tributylmethylammonium chloride solution, 75 wt. % in H2O