Merck
  • Furin inhibitor protects against neuronal cell death induced by activated NMDA receptors.
Furin inhibitor protects against neuronal cell death induced by activated NMDA receptors.
Scientific reports (2018-03-28)
Mariko Yamada, Hideki Hayashi, Moe Yuuki, Nahoko Matsushima, Bo Yuan, Norio Takagi
ABSTRACT
The proprotein convertases (PCs) act as serine proteases and are known to convert diverse precursor proteins into their active forms. Among the PCs, furin has been considered to play a crucial role not only in embryogenesis, but also in the initiation and progression of certain pathologic conditions. However, the roles played by furin with respect to neuronal cell injuries remain to be determined. An excessive influx of Ca2+ through the N-methyl-d-aspartate (NMDA) receptor has been associated with diverse neurological and neurodegenerative disorders. The aim of this study was to achieve further insight into the pathophysiologic roles of furin in cultured cortical neurons. We demonstrated that furin inhibitors dose-dependently prevented neuronal injury induced by NMDA treatment. Neuronal injury induced by NMDA treatment was attenuated by the calpain inhibitor calpeptin. And the increase observed in the activity of calpain after NMDA treatment was significantly inhibited by these furin inhibitors. Furthermore, calpain-2 activity, which was evaluated by means of the immunoblotting assay, was increased by NMDA treatment. It was noteworthy that this increased activity was almost completely inhibited by a furin inhibitor. Our findings suggested that furin is involved in NMDA-induced neuronal injury by acting upstream of calpain.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
GM 6001, InSolution, ≥95%, broad-spectrum inhibitor of MMPs
Sigma-Aldrich
Furin Inhibitor II
Sigma-Aldrich
Calpeptin, A cell-permeable calpain inhibitor (ID₅₀ = 52 nM for calpain-1; ID₅₀ = 34 nM for calpain-2; ID₅₀ = 138 nM for papain).
Sigma-Aldrich
Anti-NMDAR2A Antibody, Chemicon®, from rabbit