Merck
  • Oncoproteomic analysis reveals co-upregulation of RELA and STAT5 in carboplatin resistant ovarian carcinoma.
Oncoproteomic analysis reveals co-upregulation of RELA and STAT5 in carboplatin resistant ovarian carcinoma.
PloS one (2010-06-30)
Natini Jinawath, Chanont Vasoontara, Artit Jinawath, Xueping Fang, Kejia Zhao, Kai-Lee Yap, Tong Guo, Cheng S Lee, Weijie Wang, Brian M Balgley, Ben Davidson, Tian-Li Wang, Ie-Ming Shih
ABSTRACT
Ovarian cancer is one of the most lethal types of female malignancy. Although most patients are initially responsive to platinum-based chemotherapy, almost all develop recurrent chemoresistant tumors and succumb to their diseases. Elucidating the pathogenesis underlying drug resistance is fundamental to the development of new therapeutics, leading to improved clinical outcomes in these patients. We compared the proteomes of paired primary and recurrent post-chemotherapy ovarian high-grade serous carcinomas from nine ovarian cancer patients using CIEF/Nano-RPLC coupled with ESI-Tandem MS. As compared to their primary tumors, more than half of the recurrent tumors expressed higher levels of several proteins including CP, FN1, SYK, CD97, AIF1, WNK1, SERPINA3, APOD, URP2, STAT5B and RELA (NF-kappaB p65), which were also validated by quantitative RT-PCR. Based on shRNA screening for the upregulated genes in in vitro carboplatin-resistant cells, we found that simultaneous knockdown of RELA and STAT5B was most effective in sensitizing tumor cells for carboplatin treatment. Similarly, the NF-kappaB inhibitor, BMS-345541, and the STAT5 inhibitor, Dasatinib, significantly enhanced cell sensitivity to carboplatin. Moreover, both RELA and STAT5 are known to bind to the promoter region of Bcl-X, regulating its promoter activity. In this regard, augmented Bcl-xL expression was detected in carboplatin-resistant cells. Combined ectopic expression of RELA and STAT5B enhanced Bcl-xL promoter activity while treatment with BMS-345541 and Dasatinib decreased it. Chromatin immunoprecipitation of the Bcl-X promoter region using a STAT5 antibody showed induction of RELA and STAT5 DNA-binding segments both in naïve cells treated with a high concentration of carboplatin as well as in carboplatin-resistant cells. Proteomic analysis identified RELA and STAT5 as two major proteins associated with carboplatin resistance in ovarian tumors. Our results further showed that NF-kappaB and STAT5 inhibitor could sensitize carboplatin-resistant cells and suggest that such inhibitors can be used to benefit patients with carboplatin-resistant recurrent ovarian cancer.
Sigma-Aldrich
Anti-NFκB Antibody, p65 subunit, active subunit, clone 12H11, clone 12H11, Chemicon®, from mouse
Sigma-Aldrich
IKK Inhibitor III, BMS-345541, IKK Inhibitor III, BMS-345541, CAS 445430-58-0, is a cell-permeable, potent, selective, and allosteric site-binding inhibitor of IKK-2 (IC₅₀ = ~ 300 nM).