Skip to Content
Merck
W-13 ≥98% (HPLC)

W-13

W-13 ≥98% (HPLC)
Synonym(s):
N-(4-Aminobutyl)-5-chloro-2-naphthalenesulfonamide hydrochloride, W13
Empirical Formula (Hill Notation):
C14H17ClN2O2S · HCl
CAS Number:
Molecular Weight:
349.28

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing