
Regorafenib impurity FP-B

Synonym(s):
4-Chloro-3-(trifluoromethyl)aniline, 4-Chloro-α,α,α-trifluoro-m-toluidine, 5-Amino-2-chlorobenzotrifluoride
Linear Formula:
ClC6H3(CF3)NH2
CAS Number:
Molecular Weight:
195.57
Products
Formula Weight
Manufacturer
Melting Point (°C)
Markush Class
Markush Group
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing