![Poly(2-methyl-2-oxazoline), hydroxy terminated average Mn 5,000, PDI <1.3](/deepweb/assets/sigmaaldrich/product/structures/401/298/35bdf4e8-b6b5-4754-ad34-91135e6119fe/640/35bdf4e8-b6b5-4754-ad34-91135e6119fe.png)
Poly(2-methyl-2-oxazoline), hydroxy terminated
![Poly(2-methyl-2-oxazoline), hydroxy terminated average Mn 5,000, PDI <1.3](/deepweb/assets/sigmaaldrich/product/structures/401/298/35bdf4e8-b6b5-4754-ad34-91135e6119fe/640/35bdf4e8-b6b5-4754-ad34-91135e6119fe.png)
Synonym(s):
PMOx
Linear Formula:
CH3(C4H7NO)nOH
Products
Brand
Physical Form
Quality Segment
Markush Class
Markush Group
Polymer Composition
Polymer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing