MSTFA Reagent
Synonym(s):
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, N-Trimethylsilyl-N-methyl trifluoroacetamide, MSTFA
Linear Formula:
CF3CON(CH3)Si(CH3)3
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Quality Segment
Application
Package Size
Reaction Type
Reagent Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing