Skip to Content
Merck
Merocyanine 540 Dye content 90 %

Merocyanine 540

Merocyanine 540 Dye content 90 %
Empirical Formula (Hill Notation):
C26H32N3NaO6S2
CAS Number:
Molecular Weight:
569.67

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing