
Mac Conkey-Sorbitol ChromoSelect Agar

Synonym(s):
Mac Conkey-sorbitol Agar ChromoSelect, SMAC Agar ChromoSelect, Sorbitol MacConkey Agar ChromoSelect
Products
Product Category
Brand
pH Value
Physical Form
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing