
Isotopic standard for chlorine

Synonym(s):
Sodium chloride, Halite, NaCl
Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Quality Segment
Isotope
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing