Skip to Content
Merck
Ibuprofen Impurity N Pharmaceutical Secondary Standard; Certified Reference Material

Ibuprofen Impurity N

Ibuprofen Impurity N Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
(2RS)-2-(4-Ethylphenyl)propionic acid, (2RS)-2-(4-Ethylphenyl)propanoic acid, 4-Ethyl-α-methylbenzeneacetic acid, p-Ethylhydratropic acid
Empirical Formula (Hill Notation):
C11H14O2
CAS Number:
Molecular Weight:
178.23

Products

Brand
Formula Weight
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1