
Epicoprostanol

Synonym(s):
5β-Cholestan-3α-ol, Epicoprostanol
Empirical Formula (Hill Notation):
C27H48O
CAS Number:
Molecular Weight:
388.67
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing