
di-Potassium hydrogen phosphate

Synonym(s):
Potassium phosphate dibasic, Dipotassium hydrogenphosphate, Dipotassium phosphate, sec.-Potassium phosphate
Linear Formula:
K2HPO4
CAS Number:
Molecular Weight:
174.18
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2