Skip to Content
Merck
β-Acetyl-γ-O-hexadecyl-L-α-phosphatidylcholine hydrate ≥98%

β-Acetyl-γ-O-hexadecyl-L-α-phosphatidylcholine hydrate

β-Acetyl-γ-O-hexadecyl-L-α-phosphatidylcholine hydrate ≥98%
Synonym(s):
1-Hexadecyl-2-acetyl-sn-glycero-3-phosphocholine, 1-O-Palmityl-2-acetyl-sn-glycero-3-phosphocholine monohydrate, 2-Acetyl-1-hexadecyl-sn-glycero-3-phosphocholine, 3-sn-Phosphatidylcholine, 1-hexadecyl-2-acetyl, PAF(C16), Platelet activating factor
Empirical Formula (Hill Notation):
C26H54NO7P · xH2O
CAS Number:
Molecular Weight:
523.68 (anhydrous basis)

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing