
Bacampicillin hydrochloride

Empirical Formula (Hill Notation):
C21H27N3O7S · HCl
CAS Number:
Molecular Weight:
501.98
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing