
Amitriptyline Related Compound B

Synonym(s):
5-[3-(Dimethylamino)propyl]-10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5-ol
Empirical Formula (Hill Notation):
C20H25NO
CAS Number:
Molecular Weight:
295.42
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing