
6-Fluorescein Phosphoramidite

Synonym(s):
6-FAM-phosphoramidite
Empirical Formula (Hill Notation):
C46H58N3O10P
CAS Number:
Molecular Weight:
843.94
Products
Product Category
Brand
Color
Formula Weight
Product Line
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing