
5β-Cholestan-3α-ol

Synonym(s):
Epicoprostanol
Empirical Formula (Hill Notation):
C27H48O
CAS Number:
Molecular Weight:
388.67
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing