
4-(Methylamino)phenol sulfate

Synonym(s):
4-(Methylamino)phenol hemisulfate salt, Metol
Linear Formula:
C7H9NO · 1/2H2SO4
CAS Number:
Molecular Weight:
172.19
Products
Brand
Formula Weight
Melting Point (°C)
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing