
4-Acetamido-2,2,6,6-tetramethylpiperidine 1-oxyl

Empirical Formula (Hill Notation):
C11H21N2O2
Products
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing