
1-O-Galloyl-β-D-glucose

Synonym(s):
β-D-Glucopyranose 1-(3,4,5-trihydroxybenzoate), β-Glucogallin, 1-O-Galloyl-β-D-glucopyranose
Empirical Formula (Hill Notation):
C13H16O10
CAS Number:
Molecular Weight:
332.26
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing